Would you like to log in again?
Synapse Construction
Supplier: 3-D Molecular Designs
CSSCK-01
470337-153CS 849.5 USD InStock
470337-153
Synapse Construction
Educational Materials Biology Educational Materials AP Biology Learning Activities
Detailed modeling of a synapse.
  • Construct a synapse
  • Model neurotransmitter release and reception
  • Reusable
Engage your classroom with hands-on modeling of neuronal communication! Students can use the colorful foam pieces in the Synapse Construction Kit© to:
Discover how the resting potential of a neuron is established.
Demonstrate the propagation of an action potential down an axon.
Simulate the action of the sodium-potassium pump in resetting the resting potential.
Explore the effects of neurotransmitters acetylcholine, dopamine and GABA on a post synaptic neuron.
Model cholinergic, dopaminergic and GABAergic synapses.
Compare and contrast metabotropic and ionotropic receptors.
Analyze the impact of various substances such as nicotine, cocaine, sarin gas and propofol on neuronal signaling.
And much more!
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty