Would you like to log in again?
Ward's® Live Red Worm Culture (Eisenia Hortensis)
Supplier: VWR
Ward's®
25BRW30 BRW500
470337-638BX 230.05 USD InStock
470337-638 470336-804
Ward's® Live Red Worm Culture (Eisenia Hortensis)
Specimens Live Specimens Animals - Living Material
These common worms are also referred to as Red Wigglers.
  • Great to use in composting and fishing
  • Helps you go green, as they are great for composting
  • Chemical-free; safe for feeding to pets
Also known as euro worms, Belgian worms, Belgian reds, minis, and reds. While they are very easy to keep alive for weeks at room temperature in the cups they arrive in, we recommend if not using immediately to refrigerate between 38 to 55 °F.
Ordering information: For larger orders please give at least 2 weeks notice.
Delivery information: These products contain living materials and will ship via overnight or 2 nd day delivery. Please specify a delivery date during checkout.
Packaging: Each cup of worms come packed with sphagnum peat moss.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty