Would you like to log in again?
ReptiFauna Bioactive Desert Plants
Supplier: SEGREST FARMS
ReptiFauna
574-0009-1 574-0008-1 574-0011-1 574-0010-1
470337-640EA 11.8 USD InStock
470337-640 470337-642 470337-644 470337-646
ReptiFauna Bioactive Desert Plants
Specimens Live Specimens Plants - Living Material
Transform your terra and grow a reptile, amphibian, or invertebrate oasis with ReptiFauna plants.
  • Naturally enhance desert habitats
  • Low maintenance plants
  • Foundation of an oasis
  • Safe for animals
These bioactive plants are perfect for naturally enhancing animal habitats, and they are easy to care for, making them an excellent choice as a beginner level plant. These slow growing desert plants are great mid-ground plants that thrive in low humidity and add texture and a pop of color to pet habitats.
Delivery information: Please specify a delivery date during checkout. Live plants will ship via overnight or 2nd day delivery.
Packaging: Packed in pots with animal safe substrate.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty