Would you like to log in again?
SureSeal S™ Sterile Microcentrifuge Tubes
Supplier: MTC BIO, INC.
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
C2000-50AST C2000-50
470337-700EA 7.85 USD InStock
470337-700 470337-702
SureSeal S™ Sterile Microcentrifuge Tubes
Tubes Microcentrifuge Tubes
SureSeal S™ microcentrifuge tubes are designed for a broad range of research applications.
  • Rnase-, Dnase-, and pyrogen-free
  • Sterilized by electron beam radiation
  • Each 50 pack comes in a self-standing bag with gusseted bottom
  • 10 stop-pops supplied in every bag
  • Assorted color packs contain blue, red, purple, yellow, and green
Ideal for multiple assays including enzyme digests, DNA isolation/purification, centrifugation, incubation, and general sample/reagent storage.
SureSeal S™ tubes are manufactured under cleanroom conditions, using only BioClear™ co-polymer, an advanced polypropylene that maximizes clarity, centrifugal strength (tested up to 28000×g) and the usable temperature range (−90 to 121 °C).
The oversized tube cap (with frosted marking surface), combined with the unique ring and groove closure, enhances the security of the liquid-tight seal, while minimizing the required locking forces for a more ergonomic opening/closing of the tube caps.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty