Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Classroom Laboratory Drawer

Supplier: VWR International
UNLABSET-01WS
470337-818EA 248.85 USD InStock
470337-818
Classroom Laboratory Drawer
Educational Classroom Kits and Activities
A useful set of tools and labware for your laboratory.

  • Assortment of popular laboratory items
  • Suitable for any chemistry classroom

Laboratory Drawer™ is a convenient set of the most commonly used chemistry classroom items. Almost all of the items (except for pH paper and wax glassware marking pencil) are durable and reusable.

Delivery information: Set includes:
• Beaker set of 5, borosilicate glass, (1 each 50, 100, 250, 600 and 100 ml)
• Clamp, 3-prong extension with boss head, die-cast zinc with chrome plating, 2.25" jaw opening, (2)
• Crucible tongs, 9”, plated steel
• Crucible, porcelain, wide-form with cover, 50 ml capacity
• Erlenmeyer flask set of 5, borosilicate glass, (1 each 50, 125, 250, 500 and 1000 ml)
• Funnel, utility, polypropylene, 25 ml capacity, 30 mm stem length
• Graduated cylinder, borosilicate glass, 5 ml capacity
• Graduated cylinder, borosilicate glass, 10 ml capacity
• Graduated cylinder, borosilicate glass, 25 ml capacity
• Laboratory tray, polypropylene
• Laboratory scoop, stainless steel, 6.5" length
• Pencil, wax marking for glassware
• pH paper, wide range, 100 strips
• Safety goggles, chemical splash, PVC with polycarbonate lens, ANSI Z87.1-2020 certified
• Spatula, stainless steel, 6" length, both ends flat
• Stirring rod, flint glass, 8" long, 5 mm diameter, (2)
• Support ring with bosshead clamp, cast iron, 3" diameter
• Support stand with rod, stamped steel, black enamel finish, 5×8" base with 20" rod
• Test tubes with rack, 12 tubes with 24 ml capacity and rubber stoppers, polypropylene unassembled rack
• Thermometer, glass, 12", partial immersion, −20 to 110 °C
• Watch glass, flint glass, 50 mm diameter, (2)
• Well plate, 12-well, polystyrene
• Wire gauze, plain, 5×5", iron 24SWG/10 mesh with folded edges

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty