Would you like to log in again?
Wards® Solid Fuel Chemical Demonstration Kit
Supplier: ALDON CORP SE
Wards®
IS7025
470337-906EA 25.75 USD InStock
470337-906
Wards® Solid Fuel Chemical Demonstration Kit
Educational Materials Chemistry Educational Materials Thermochemistry/Kinetics Learning Activities
Explore a solid that can be ignited with a match!
  • Contains enough materials for 2 demonstrations
Two solutions are poured together in a beaker and form a gel. The gel can be ignited with a match. Commercially this gel is sold as Sterno®.
Delivery information: Includes 2×25 ml Ethyl alcohol, denatured; 2×25 ml Calcium acetate solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty