Would you like to log in again?
Wards® Ampholytes Demonstration
Supplier: ALDON CORP SE
Wards®
IS7003
470337-908EA 25.5 USD InStock
470337-908
Wards® Ampholytes Demonstration
Educational Materials Chemistry Educational Materials Acids and Bases Learning Activities
Discover the dual reactivity of ampholytes!
  • React a single substance with both an acid and a base
Ampholytes are compounds which react with both acids and bases. When Zinc chloride is reacted with sodium hydroxide the soluble zincate ion is formed. When Zinc hydroxide is reacted with hydrochloric acid, the soluble zinc chloride and water are formed.
Delivery information: Includes 1×100 ml Zinc chloride, 0.5 M solution; 1×250 ml Sodium hydroxide, 1.0 M solution; 1×250 ml Hydrochloric acid, 1.0 M solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty