Would you like to log in again?
Wards® Formation of Tin Wool Demo
Supplier: ALDON CORP SE
Wards®
IS7032
470337-912EA 64 USD InStock
470337-912
Wards® Formation of Tin Wool Demo
Educational Materials Chemistry Educational Materials Electrochemistry Learning Activities
Form tin wool through a single replacement reaction.
  • Includes enough materials for 7 demonstrations
This is a single replacement reaction in which the zinc metal dissolves in solution and tin metal is formed.
Delivery information: Includes 2×700 ml Tin (II) chloride, 1.0 M solution; 1×140 g Zinc metal mossy.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty