Would you like to log in again?
Wards® Exothermic Reactions Chemical Demonstration Kit
Supplier: ALDON CORP SE
Wards®
IS7004
470337-914EA 27 USD InStock
470337-914
Wards® Exothermic Reactions Chemical Demonstration Kit
Educational Materials Chemistry Educational Materials Thermochemistry/Kinetics Learning Activities
Study the process of exothermic reactions.
  • The reaction is safely carried out in a plastic bag
  • The students can pass the bag around to feel the result of the reaction
Exothermic reactions are reactions that release heat. Demonstrate a real life example of one of these chemical reactions to your students in a hands-on way!
Delivery information: 5 bags of the following mixture: 25 g Iron metal powder; 1 g Sodium chloride crystals; 5 g Calcium chloride flakes; 2 tbsp Vermiculite.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty