Would you like to log in again?
Wards® Exothermic/Endothermic Combination Demonstration
Supplier: ALDON CORP SE
Wards®
IS7006
470337-924EA 44.95 USD InStock
470337-924
Wards® Exothermic/Endothermic Combination Demonstration
Educational Materials Chemistry Educational Materials Thermochemistry/Kinetics Learning Activities
Study endothermic and exothermic reactions with this hands-on demonstration.
  • Contains enough materials for 5 demonstrations
Study both processes of endothermic and exothermic reactions. Reactions are safely carried out in plastic bags. The students can pass the bags around to feel the results of the reactions.
Delivery information: 5 bags of the following mixture: 25 g Iron metal powder; 1 g Sodium chloride crystals; 5 g Calcium chloride flakes; 2 tbsp Vermiculite; 8 package×2 tablets Sodium bicarbonate/Citric acid tablets; 15 Plastic cups; 15 Stirring sticks; 15 Bags containing: 25 g Ammonium nitrate, 1 tbsp Vermiculite.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty