Would you like to log in again?
Wards® Autocatalysis Chemical Demonstration Kit
Supplier: ALDON CORP SE
Wards®
IS7002
470337-930EA 28.5 USD InStock
470337-930
Wards® Autocatalysis Chemical Demonstration Kit
Educational Materials Chemistry Educational Materials Thermochemistry/Kinetics Learning Activities
Witness a reaction that catalyzes itself!
  • Contains enough materials for 5 demonstrations
Catalysis is a basic principle of chemistry and biochemistry. Some reactions are autocatylic; the product of the reaction actually catalyzes further reactions. The reaction is initiated with acid and produces acid to continue the reaction.
Delivery information: Include 1×5 ml Toluidine Blue, 0.5% solution; 1×30 ml Sulfuric acid, 3.0 M solution; 1×250 ml Potassium chlorate/Sodium sulfite solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty