Would you like to log in again?
Wards® Endothermic Reactions Demonstration
Supplier: ALDON CORP SE
Wards®
IS7005
470337-934EA 27.05 USD InStock
470337-934
Wards® Endothermic Reactions Demonstration
Educational Materials Chemistry Educational Materials Thermochemistry/Kinetics Learning Activities
Study the process of endothermic reactions.
  • Contains enough materials for 5 demonstrations
Study the process of endothermic reactions. The reaction is safely carried out in a plastic bag. The students can pass the bag around to feel the result of the reaction.
Delivery information: Includes 8 package×2 tablets Sodium bicarbonate/Citric acid tablets; 15 Plastic cups; 15 Stirring sticks; 15 Bags containing: 25g Ammonium nitrate, 1 tbsp Vermiculite.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty