Would you like to log in again?
Wards® Oxidation of Glycerin Chemical Demonstration
Supplier: ALDON CORP SE
Wards®
IS7028
470337-940EA 21.75 USD InStock
470337-940
Wards® Oxidation of Glycerin Chemical Demonstration
Educational Materials Chemistry Educational Materials Electrochemistry Learning Activities
Discover an exothermic reaction in which violet flames are produced!
  • Contains enough materials for 3 demonstrations
A small amount of potassium permanganate is placed in a petri dish. A few drops of glycerin are added and after a few seconds, a puff of smoke and violet flames are produced.
Delivery information: Includes 1×30 g Potassium permanganate; 1×25 ml Glycerin.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty