Would you like to log in again?
Wards® LeChatlier's Principle Demonstration
Supplier: ALDON CORP SE
Wards®
IS7034
470337-942EA 32 USD InStock
470337-942
Wards® LeChatlier's Principle Demonstration
Educational Materials Chemistry Educational Materials Thermochemistry/Kinetics Learning Activities
Find out how equilibrium is reestablished when system at equilibrium is disturbed!
  • Use an overhead projector to study the reaction
Le Chatelier’s principle states that if a system at equilibrium is disturbed by a change in reaction conditions, the system will shift to counteract the change and reestablish an equilibrium. The color changes in this demonstration allow these shifts to be observed.
Delivery information: Includes 1×140 ml Potassium thiocyanate, 0.002 M solution; 1×10 ml Iron (III) Nitrate, 0.2 M solution; 1×5 g Potassium thiocyanate, crystals; 1×5 g Sodium phosphate, monobasic.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty