Would you like to log in again?
Wards® Synthetic Rubber Chemical Demonstration Kit
Supplier: ALDON CORP SE
Wards®
IS7008
470337-944EA 36.95 USD InStock
470337-944
Wards® Synthetic Rubber Chemical Demonstration Kit
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Discover how the first synthetic rubber in the United States was made!
  • Contains enough materials for 4 demonstrations
Show how the first synthetic rubber was made in United States by a reaction of sodium polysulfide with ethylene chloride. This produces a simple condensation polymer consisting of repeating units of ethane and polysulfide.
Delivery information: Includes 4×25 ml Sodium polysulfide solution; 1×25 ml Ethylene dichloride.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty