Would you like to log in again?
Wards® Water to Wine Chemical Demonstration Kit
Supplier: ALDON CORP SE
Wards®
IS7035
470337-948EA 44.25 USD InStock
470337-948
Wards® Water to Wine Chemical Demonstration Kit
Educational Materials Chemistry Educational Materials Acids and Bases Learning Activities
Study acid base indicators and complex ions by mixing colored chemical solutions.
  • Includes enough materials for 7 demonstrations
Study acid base indicators and complex ions. Mixing three colored chemical solutions results in another wine colored solution. Adding the fourth chemical of sodium fluoride turns the solution clear.
Delivery information: Includes 1×10 ml Potassium thiocyanate, 1 M solution; 1×15 ml Thymolphthalein, 0.04% solution; 1×15 ml Phenolphthalein, 0.5% solution; 1×5 g Iron (III) chloride, Hexahydrate; 3×25 ml Sodium Fluoride, 1 M solution; 1×10 ml Hydrochloric acid, 32%; 1×10 ml Ammonium hydroxide, 14.8 M solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty