Would you like to log in again?
Wards® Formation of Silicate (Red and White Precipitates) Chemical Demonstration
Supplier: ALDON CORP SE
Wards®
IS7011
470337-950EA 23.5 USD InStock
470337-950
Wards® Formation of Silicate (Red and White Precipitates) Chemical Demonstration
Educational Materials Chemistry Educational Materials Ionic Properties Learning Activities
Explore the solubility of silicates from monovalent and divalent ions.
  • Contains enough materials for 5 demonstrations
Show how silicates of divalent ions are insoluble in water whereas silicates of monovalent ions are soluble.
Delivery information: Includes 50 ml Calcium chloride, 1.0 M solution; 50 ml Calcium chloride, 1.0 M solution with Phenolphthalein; 200 ml Sodium silicate, 15% Aqueous solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty