Would you like to log in again?
Wards® Negative Coefficient of Solubility Chemical Demo
Supplier: ALDON CORP SE
Wards®
IS7016
470337-954EA 21.25 USD InStock
470337-954
Wards® Negative Coefficient of Solubility Chemical Demo
Educational Materials Chemistry Educational Materials Mixtures and Solutions Learning Activities
Explain how there are exceptions to the rules of chemistry.
  • Demonstrates reversible precipitation
Heat a solution and watch it form a precipitate. Watch it go back into solution as it cools.
Delivery information: 1×500 ml Calcium acetate, Saturated solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty