Would you like to log in again?
Microslide Viewer
Supplier: SUPERTEK SCIENTIFIC LLC
T001
470338-118EA 23.75 USD InStock
470338-118
Microslide Viewer
Microscopes
The Microslide Viewer is specialty designed instrument for viewing Microslides®.
  • Light weight and easy to use!
  • No electricity or batteries required
Simply insert the slide and focus. The viewer brings 5X magnification allowing every image to be seen exactly as if observed through a microscope under ideal conditions. The white reflector provides sufficient light for a bright image in the normal light of a classroom. The viewer does not require electricity or batteries so it can also be used outdoors. 9″ high. For budding scientists of all ages!
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty