Would you like to log in again?
Discover Simulation Headwall Packages
Supplier: DIAMEDICAL SE
HW020800 HW020802 HW020804 HW020803 HW020805
470338-468EA 5919.79 USD InStock
470338-468 470338-358 470338-424 470338-464 470338-454
Discover Simulation Headwall Packages
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Simulation headwall with air.
  • Multiple sizes available
  • Wall mounted; mounting strip included
  • Nurse call facade with code blue button and hand pendant
  • Includes silent air compressor
  • Includes complete accessories package
The Functioning Wall Mounted Discover Headwall Package simulates oxygen, air, and suction with one source of air by connecting to an external air compressor. This solution is thousands of dollars less than a traditional centralized system with air, oxygen, and vacuum.
Please see product resources for contents.
Ordering information: Default color is off white. Other colors, sizes, and configurations are available. Please consult with your sales representative for more information.
Caution: This is a non-functional headwall intended for simulation training.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty