Would you like to log in again?
SimLabSolutions Injection Trainer
Supplier: DIAMEDICAL SE
MS153901 MS153902
470338-474EA 47.49 USD InStock
470338-474 470338-428
SimLabSolutions Injection Trainer
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Easily transportable injection training.
  • Real feel for skin puncture resistance
  • Self-sealing
  • Foam core may be injected with water
  • Drains easily and dries without deteriorating
  • Removable top layer is suitable for practicing intradermal injections
The SimLabSolutions injection trainers are a convenient and easy way to practice different injection techniques. The smaller pad is a perfect size for an individual student, while the larger size has a higher surface area for longer classroom durability. The container protects the pad between uses, as well as acting as a shield from accidental over sticks. The pad accommodates both dry and wet practice. The foam core is easily drained and dried.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty