Would you like to log in again?
SimLabSolutions Suture Training
Supplier: DIAMEDICAL SE
MS153905 MS153907 MS153906
470338-462EA 77.19 USD InStock
470338-462 470338-480 470338-390
SimLabSolutions Suture Training
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Multi-layer suture training.
  • Trainees can repeatedly suture incision sites until skills are mastered
  • Non-slip pad beneath the dermis eliminates the need for suturing cradles
  • Practiskin™ will hold superficial and deep sutures without tearing
  • Epidermis and dermis have realistic look and feel
If it’s the full kit, the pad and cradle, or just the skin pad, this practiSkin is a great solution for suture training. Each pad is a multi-layer training pad, allowing students to practice suturing at different tissue layers. The non-skid base holds the pad securely without the need for a base. The cradle allows students to practice on a rounded surface, providing more tension required for wound closure.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty