Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ward's® Mini Lab Notebook

Supplier: Heathrow Scientific
Ward's®
120893W
470338-494CS 450.19 USD InStock
470338-494
Ward's® Mini Lab Notebook
Notebooks
This mini lab notebook is a compact solution to traditional lab notebooks.

  • Quality Smyth Sewn binding holds pages securely and lies flat when opened
  • User data, table of contents, notes section and conversion tables are included in every book
  • Durable hardbound, water-resistant cover
  • Marker Ribbon to keep your place
  • Dimensions: 4.9×8" (12.3×20.3 cm)
  • Grid: 5×5 mm grid size
  • Color: Black

The Ward's laboratory notebook is ideal for a variety of applications such as recording research data, experimental procedures and design, development ideas, and more. Use this notebook to keep information safe and well organized.

Made of acid-free paper, pages are numbered and include space for title, date, project number, book number, date, and witness signature.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty