Would you like to log in again?
HOBO Weather Station Kits
Supplier: Avantor
SP-610
470338-514EA 2412.19 USD InStock
470338-514 470338-518 470348-910 470338-516
HOBO Weather Station Kits
Weather Stations
HOBO research-grade cellular HOBO weather station kits offer accurate, reliable performance - even in the harshest outdoor climates.
  • Wide coverage with 900 MHz wireless mesh technology provides a stronger, more reliable signal that can communicate through vegetation
  • Scalable, with up to 50 wireless sensors streaming back to a central, cloud-based weather station
  • Remote access to data and current conditions with a customizable dashboard for analysis
  • Alarm notifications when conditions reach user-set thresholds
  • Data access through Onset's cloud-based HOBOlink software
  • Rain gauge sensor - S-RGA-M002
  • Temp/RH sensor accuracy - S-THC-M002
The award-winning HOBOnet field monitoring system from Onset provides a cost-effective and scalable solution for web-enabled monitoring of field conditions for applications such as crop management, research, and greenhouse operations.
Choose either the easy-to-deploy HOBO MicroRX station or the highly flexible HOBO RX3000 station as the basis of your HOBOnet system. Easily configure systems for your specific applications with HOBOnet wireless sensors or Onset's plug-and-play sensors.
Ordering information: Included in basic kit: RX2100 data logger, data plan (must be renewed annually), Temp/RH sensor with solar radiation shield, wind speed and direction sensor, rain gauge sensor, 2m tripod mounting kit. Included in intermediate kit: RX3004 data logger, data plan (must be renewed annually), solar panel (5 W), Temp/RH sensor with radiation shield, wind speed and direction set, rain guage, 2m tripod mounting kit with full cross arm. Included in intermediate kit: RX3004 data Logger, data plan (must be renewed annually), solar panel (5 W), Temp/RH sensor with radiation shield, wind speed and direction sensor set, rain gauge, barometric pressure sensor, solar radiation sensor, 2 m tripod mounting kit with full cross arm and light sensor.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty