Would you like to log in again?
Foam City - The Catalytic Decomposition of Hydrogen Peroxide
Supplier: ALDON CORP SE
IS7019
470338-542EA 25.6 USD InStock
470338-542
Foam City - The Catalytic Decomposition of Hydrogen Peroxide
Educational Materials Chemistry Educational Materials Mixtures and Solutions Learning Activities
A large quantity of foam is produced when detergent and potassium iodide are added to hydrogen peroxide.
  • Understand and demonstrate a catalytic decomposition reaction
  • Aligned to NGSS practices
  • Cross cutting concepts: patterns
  • Disciplinary core ideas: PS1.A, PS1.B
  • Learn engineering practices: analyzing and interpreting data
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty