Would you like to log in again?
Synthesis of Rayon
Supplier: ALDON CORP SE
IS7001
470338-552EA 31.85 USD InStock
470338-552
Synthesis of Rayon
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Demonstrate polymer chemistry.
  • Understand and demonstrate polymer chemistry through a synthesis reaction
  • Aligned to NGSS practices
  • Cross cutting concepts: patterns, structure and function
  • Disciplinary core ideas: PS1.A, PS1.B
  • Learn engineering practices: analyzing and interpreting data, obtaining, evaluating, and communicating information
Newsprint is dissolved in a solution of ammonium hydroxide and cupric sulfate. The solution is then added to a sulfuric acid solution and dark strands are formed.
Caution: WARNING: This product can expose you to chemicals including strong inorganic acid mists containing sulfuric acid, which are known to the state of California to cause cancer. For more information go to www.P65Warnings.ca.gov
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty