Would you like to log in again?
Forming Red, White, & Blue
Supplier: ALDON CORP SE
IS7036
470338-566EA 34.65 USD InStock
470338-566
Forming Red, White, & Blue
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Introduce different chemical reaction types through a colorful demonstration.
  • Understand and demonstrate different chemical reaction types
  • Aligned to NGSS practices
  • Cross cutting concepts: patterns
  • Disciplinary core ideas: PS1.A, PS1.B
  • Learn engineering practices: analyzing and interpreting data
Introduce chemical reaction types. This is a patriotic demonstration that illustrates complex ion formation, double replacement while introducing chemical reaction types.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty