Would you like to log in again?
Water to Wine
Supplier: ALDON CORP SE
IS7035
470338-576EA 44.25 USD InStock
470338-576
Water to Wine
Educational Materials Chemistry Educational Materials Acids and Bases Learning Activities
Understand acid base indicators and complex ions.
  • Understand and demonstrate acid-base chemistry and complex ions
  • Aligned to NGSS Practices
  • Cross Cutting Concepts: Patterns
  • Disciplinary Core Ideas: PS1.A, PS1.B
  • Learn Engineering Practices: Analyzing and interpreting data
Study acid base indicators and complex ions. Mixing three colored chemical solutions results in another wine colored solution. Adding the fourth chemical of sodium fluoride turns the solution clear.
Caution: WARNING: This product can expose you to chemicals including methanol/methyl isobutyl ketone and phenolphthalein, which are known to the state of California to cause cancer. For more information go to www.P65Warnings.ca.gov
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty