Would you like to log in again?
Underwater Fireworks - The Reaction of Acetylene and Chlorine
Supplier: ALDON CORP SE
IS7029
470338-584EA 26.55 USD InStock
470338-584
Underwater Fireworks - The Reaction of Acetylene and Chlorine
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Create fireworks by conducting a chemical reaction underwater.
  • Understand and demonstrate an underwater chemical reaction
  • Aligned to NGSS practices
Calcium carbide is placed in a cylinder containing water. A reaction occurs producing acetylene gas bubbles. A plastic tube from a chlorine gas generator is placed in the cylinder. As the bubbles of acetylene and chlorine collide flashes of light are produced.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty