Would you like to log in again?
Petri Dishes
Supplier: GSC INTERNATIONAL SE
1500-9-10 1500-4-CS 1500-3-CS 1500-10-10 1500-7-10 1500-8-10
470338-702CS 71.95 USD InStock
470338-702 470338-724 470338-664 470338-642 470338-694 470338-626
Petri Dishes
Culture Dishes Petri Dishes
In a science lab or classroom, a petri dish is used to culture different types of cells, including bacteria and molds.
  • Durable and resistant to cracks and scratches
  • Not autoclavable
Get the reliable Petri dish you need for your scientific work. Crafted from high-quality soda lime glass, our Petri dishes are built to last, resistant to cracks and scratches for dependable performance. While they may not be autoclavable, the two-piece design with a lid and bottom dish provides ample space and easy access to protect your cell cultures.
Order this case of 100 Petri dishes now and stock up on the essential tools for your experiments and research. With this bulk supply, you'll save time and money, ensuring you have the precision glassware to support your scientific endeavors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty