Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ultralast Green Rechargeable Batteries
Supplier: DANTONA INDUSTRIES INC SE
UL357 ULGED4AAA UL2032 ULGED4AA
470338-834CS 26.3 USD InStock
470338-834 470338-828 470338-826 470338-836
Ultralast Green Rechargeable Batteries
Batteries
Performs exactly like the batteries you're used to, so there's no compromise in getting greener.
  • Ready to use right out of the pack, just like regular batteries
  • Keep a charge for up to 6+ months in low power consumption devices such as remote controls, clocks, flashlights, and other common household items
  • Holds 80% of charge after 12 months when not in use
  • Can be charged as many as 500 times, or up to three years
  • One rechargeable battery can keep up to 1000 disposable alkaline batteries out of landfills
  • Can be recycled through the Rechargeable Battery Recycling Corporation (RBRC) program
  • Packaged in eco-friendly materials, including post-consumer recycled cardboard and recyclable PET plastic
By replacing single-use batteries with Everyday Rechargeables, users can save money and prevent toxic landfill as they are reused over and over again. Using Ultralast Green batteries is an easy way to take a greener step every day.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty