Would you like to log in again?
UniStore™ Reagent Bottles, Narrow Mouth, HDPE
Supplier: United Scientific Supplies
UniStore™
33403-BULK 33402 33418 33416 33403 33404 33401-BULK 33401 33405 33404-BULK 33402-BULK 33405-BULK
470338-866EA 284.9 USD InStock
470338-866 76263-122 470346-156 76263-124 470339-436 470346-166 76263-120 470339-916 470339-088 76263-126 470339-318 76263-128
UniStore™ Reagent Bottles, Narrow Mouth, HDPE
Bottles Plastic Bottles
Bottles are for general laboratory use and for storing, shipping, and packing liquids.
  • Translucent
  • Pliable
UniStore™ narrow mouth HDPE reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant. Excellent chemical resistance to most acids, bases, and alcohols.
Ordering information: Polypropylene caps are included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty