Would you like to log in again?
Child Size Safety Goggles
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
UNGOGGLE-S
470338-870EA 3.25 USD InStock
470338-870
Child Size Safety Goggles
Goggles
Small sized safety goggles to protect the eyes.
  • Highly durable with polycarbonate lens made for excellent visibility and protection
  • Single-lens design for a clear and unobstructed view
  • Adjustable comfort elastic headband
  • Safe eye protection for lab work, chemistry, woodworking, mechanics, and more
Durable plastic goggles with polycarbonate lenses. Flexible body increases wearing comfort. The 5" lens size is smaller than standard goggles to provide a better fit for students. Meets ANSI Z87.1+ safety standards.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty