Would you like to log in again?
Silicone Stoppers, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
UNSST7 UNSST2 UNSST3 UNSST0 UNSST5 UNSST5A UNSST7A UNSST4 UNSST6A UNSST8 UNSST6 UNSST00 UNSST1
470340-104EA 4.5 USD InStock
470340-104 470339-106 470339-228 470340-262 470339-748 470340-132 470340-000 470339-738 470338-900 470338-876 470339-174 470339-790 470339-230
Silicone Stoppers, United Scientific Supplies
Stoppers Silicone Stoppers
Chemical resistant solid silicone stoppers.
  • Sturdy non reactive silicon
Silicone stoppers are particularly useful due to their inert and nonreactive nature. It will not impair the growth of tissue cultures and resistant to most chemical acids and solvents. Stoppers can be steam sterilized at 15 lbs. pressure. Broad working temperature range of –62 to +232 °C (–80 to +450 °F).
Durometer hardness ~50.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty