Would you like to log in again?
Conductivity Of Solutions Apparatus
Supplier: United Scientific Supplies
CSA001
470338-878EA 21 USD InStock
470338-878
Conductivity Of Solutions Apparatus
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Demonstrate the electrical conductivity of solutions.
  • 3 V conductivity tester demonstrates the electrical properties of various liquids
  • Includes activity guide with step-by-step instructions
This 3 V conductivity tester demonstrates the electrical properties of various liquids. The item consists of a molded plastic lamp socket mounted on a base with two binding posts for connecting a wire cord. It comes complete with a bulb and glass tumbler. Includes activity guide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty