Would you like to log in again?
Optics Set
Supplier: United Scientific Supplies
OPSET1
470338-888EA 29.1 USD InStock
470338-888
Optics Set
Educational Materials Physics Educational Materials Optics Learning Activities
Investigate converging and diverging lenses and mirrors.
  • Includes a storage case
  • Concave and convex components
Includes two acrylic lenses, one each of double convex and double concave and three mirrors, one each of convex, concave and plane. Packed in a cushioned wood storage box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty