Would you like to log in again?
United Scientific Student Force Table
Supplier: United Scientific Supplies
FTE001
470339-000EA 230 USD InStock
470339-000
United Scientific Student Force Table
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
Illustrate force vectors and resultants.
  • Complete student force table set features a dual 0.5 degree scale
  • Lightweight, durable table top, with all-metal tripod stand
  • Table diameter is 40.6 cm (16")
  • Includes activity guide
This student force table is a complete set that features a dual scale; an outer scale for accurate pulley positioning, and an inner scale for orienting a tracing paper overlay. Set includes four complete weight sets (each with brass slotted weights, 2×5 g, 4×10 g, 4×20 g, and 20 g brass weight hanger), four pulleys with clamps and two sets of strings that allow students to investigate the equilibrium of either three or four forces. A 16" diameter lightweight, durable table top and sturdy cast iron support allow both ease of use and accuracy. Includes activity guide.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty