Would you like to log in again?
Resonance Apparatus
Supplier: United Scientific Supplies
RSAPP01
470339-036EA 111.8 USD InStock
470339-036
Resonance Apparatus
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Investigate the wavelength of sound on an open end tube.
  • 99 cm working tube length
  • Metric scale along the length of the tube
  • Aligned with NGSS Standards
The Resonance Apparatus allows students to investigate the resonant behavior of an enclosed air column when it is stimulated by an external sound source. The air column lengths for multiple resonances at various frequencies can be found and the resulting data can be used to determine the speed of sound. A cylindrical acrylic tube is mounted vertically on a stand base and rod. The tube is open at the top and closed at the bottom by a rubber stopper with a hose barb.
A scale is printed along the length of the tube. Flexible tubing connects the bottom of the tube to a large reservoir. The reservoir and tube are partially filled with water, creating an air space in the tube above the water level. The length of the space can be varied by adjusting the height of the reservoir on the support rod.
A suitable sound source, which can be a tuning fork (not included) or a tone generator (not included), is held close the mouth of the tube while the length of the air column is varied. Points of resonance where the sound in the tube becomes louder can be heard and measured. The sound level may also be recorded for later analysis using probe ware.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty