Would you like to log in again?
Filtering Kit
Supplier: United Scientific Supplies
FLTKIT
470339-072EA 94.35 USD InStock
470339-072
Filtering Kit
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
Compact kit for vacuum filtration.
  • Complete kit, including hand vacuum pump, enables faster liquid filtration
  • Hand vacuum pump with gauge also available separately (HVP001)
This easy-to-use set serves as a faster alternative to gravy-based liquid filtration.
Kit contents: Hand vacuum pump with gauge (HVP001), 500 ml filtering flask, 90 mm porcelain Buchner funnel, Pack of 100 filter papers (90 mm diameter), Rubber stopper, Vacuum hose, and Activity guide.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty