Would you like to log in again?
United Scientific™ Petri Dishes, Polystyrene, United Scientific Supplies
Supplier: United Scientific Supplies
K1004-J-PK/500 K1006-PK/650 K1003-PK/500 K1001-PK/180 K1007-PK/500
470339-446EA 108.1 USD InStock
470339-446 470339-078 470340-170 470339-620 470338-872
United Scientific™ Petri Dishes, Polystyrene, United Scientific Supplies
Culture Dishes Petri Dishes
Sterile and disposable petri dishes.
  • Mold in grid on lid
  • Excellent optical clarity
  • Disposable polystyrene
  • Sterilized
Disposable polystyrene petri dishes offer optical clarity and strength at economical prices. 65 mm dish includes a molded-in grid on the lid for easy colony counting. Not Slippable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty