Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Cryo Racks, United Scientific Supplies
Supplier: United Scientific Supplies
66302 66303 66301
470339-522EA 52.9 USD InStock
470339-522 470339-262 470340-044
Cryo Racks, United Scientific Supplies
Racks Cryogenic Vial Racks
Strong polycarbonate boxes for storage of cryogenic vials.
  • Autoclavable
  • Polycarbonate
  • Temperatures range: –196 to +121 °C
Manufactured of strong polycarbonate, these boxes are ideal for compact storage of 2.0 or 5.0 ml cryogenic vials. Boxes can be used at temperatures from –196 to 121 °C and are autoclavable. Transparent lids have a numbered grid for sample identification. Frosted surface area is suitable for writing with markers. Dimensions of cryo boxes are designed to be compatible with vertical, horizontal, and drawer style freezer racks. 66301 includes two red boxes, one blue box, and one yellow box. 66302 includes two red boxes and two blue boxes. 66303 includes an assortment of red, yellow, and blue colored boxes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty