Would you like to log in again?
3-Prong Heavy-Duty Extension Clamp with Stainless Steel Rod, United Scientific Supplies
Supplier: United Scientific Supplies
CLHD03-VWR CLHD01 CLHD02
470339-272EA 35.35 USD InStock
470339-272 470338-906 76293-362
3-Prong Heavy-Duty Extension Clamp with Stainless Steel Rod, United Scientific Supplies
Clamps General Purpose Clamps
These extension clamps features heavy-duty electroplated zinc alloy construction and dual prong adjustments. Clamps have a maximum temperature use of 80 °C.
  • PVC coated jaws
  • Stainless steel rod
6.75" extension clamp: The 4" long extension rod is constructed of stainless steel. Prongs open to about 1.75", have a recommended maximum grip size of about 1.5", and feature double keyed spring-type adjustment screws. Prongs are covered with non-slip PVC sleeves and includes fiberglass sleeves.
9" extension clamp: The 5" long extension rod is constructed of stainless steel. Prongs open to about 2.75", have a recommended maximum grip size of about 2.5", and feature double keyed spring-type adjustment screws. Prongs are covered with non-slip PVC sleeve and includes fiberglass sleeves.
10.75" extension clamp: The 5" long extension rod is constructed of stainless steel. Prongs open to about 4", have a recommended maximum grip size of about 3.25", and feature double keyed spring-type adjustment screws. Prongs are covered with non-slip PVC sleeves.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty