Would you like to log in again?
Heat Conductometer
Supplier: United Scientific Supplies
CNDM01
470339-316EA 24.25 USD InStock
470339-316
Heat Conductometer
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Demonstrate the speed of heat conduction in five different metals.
  • Apparatus to compare the thermal conductivities of aluminum, brass, steel, stainless steel, and copper
  • Five radial spokes attached to a brass hub, with notches in each rod to hold a wax pellet
  • Each spoke is marked with the metal name on the hub
  • Unit comes assembled; includes a supply of wax and an activity guide
This item is used for comparing the thermal conductivities of aluminum, brass, steel, stainless steel, and copper. The five radial spokes are attached to a brass hub. Each rod is marked with the metal name on the hub. There are notches in each rod to hold a wax pellet.
The unit comes assembled and with a supply of wax. Includes activity guide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty