Would you like to log in again?
Radiometer
Supplier: United Scientific Supplies
RADIO1
470339-324EA 15.15 USD InStock
470339-324
Radiometer
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Classic experiment of heat creating motion.
  • Ideal for classroom demonstrations of electromagnetic radiation
  • Glass bulb with much of the air removed; low-friction spindle inside holds a rotor with 4 vanes
  • When exposed to light, the vanes turn due to differential heating
Radiometer made from a glass bulb, from which much of the air has been removed. Inside the bulb, on a low-friction spindle, is a rotor with four vanes. Each vane has one black side and one silver side; one black vane features a white dot in order to count rotations easily. When exposed to sunlight or artificial light, the vanes turn with no apparent motive power. This effect is due to differential heating of the black and silver vanes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty