Would you like to log in again?
PP Graduated Cylinder Sets with Hex Base, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CPSET7-H CPSET5
470339-350EA 47.75 USD InStock
470339-350 470339-396
PP Graduated Cylinder Sets with Hex Base, United Scientific Supplies
Cylinders Measuring Cylinders
Convenient measuring cylinder set.
  • Graduated polypropylene cylinders
  • Feature hexagonal base for excellent stability
A convenient set of seven graduated polypropylene cylinders in sizes 10 ml, 25 ml, 50 ml, 100 ml, 250 ml, 500 ml, and 1000 ml. Also available as a set of five in sizes 10 ml, 50 ml, 100 ml, 250 ml and 500 ml.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty