Would you like to log in again?
UniStore™ Reagent Bottles, Wide Mouth, HDPE
Supplier: United Scientific Supplies
UniStore™
33410 33410-BULK 33408 33407 33409-BULK 33441 33409 33412 33414 33408-BULK 33407-BULK 33406 33406-BULK
76263-100PK 26.9 USD InStock
76263-100 76263-102 470339-008 470349-760 76263-098 76263-104 76263-106 470340-098 470349-784 470339-570 470349-772 470339-130 470339-472
UniStore™ Reagent Bottles, Wide Mouth, HDPE
Bottles Plastic Bottles
Bottles are for general laboratory use and for storing, shipping, and packing liquids.
  • Translucent
  • Pliable
UniStore™ wide mouth HDPE reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant. Excellent chemical resistance to most acids, bases, and alcohols.
Ordering information: Polypropylene caps are included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty