Would you like to log in again?
Mini Pipettes, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PMP1000 PMP100 PMP020 PMP500 PMP010 PMP050 PMP200 PMP025 PMP005 PMP250
470339-478EA 29.45 USD InStock
470339-478 470339-578 470340-116 470340-280 470339-378 470339-554 470339-140 470339-022 470340-264 470339-370
Mini Pipettes, United Scientific Supplies
Pipettes Repetitive Pipettors
Mini pipettes for reverse pipetting and repetitive pipetting.
  • Compact, yet ergonomic design that handles like a syringe; can be used for reverse pipetting and repetitive pipetting
  • Two-step plunger facilitates high accuracy and precision to the last drop
  • Fully autoclavable
These mini pipettes include a two step plunger that facilitates super blowout from the tip (last drop dispensing), resulting in very high accuracy and precision. This allows the mini pipette to be used for reverse pipetting and repetitive pipetting.
The tip cone of the 5 μl, 10 μl, and 20 μl mini pipettes is designed to accept both standard 200 μl tips or ultra micro tips up to 20 μl. Use of ultra micro tips for volumes up to 20 μl enhances accuracy and precision. Can be used for reverse pipetting and repetitive pipetting.
These pipettes provide an optimum length of 130mm for user comfort during pipetting, and are fully autoclavable at 121°C, 15 psi for 10 to 15 minutes. Plungers are color coded for different volumes for easy identification in the lab.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty