Would you like to log in again?
Multi-Use Bench Pulley
Supplier: United Scientific Supplies
PULB01
470339-480EA 33.6 USD InStock
470339-480
Multi-Use Bench Pulley
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
High-quality pulley can be mounted on a support rod or table.
  • Fits table tops up to 1.75" (4.5 cm) thick
  • Constructed of heavy-duty cast aluminum support frame
  • Mounted on heavy duty, low-friction cone bearings
  • Can be mounted on a support stand rod
High-quality bench pulley that can be mounted on a support stand rod. Constructed of a heavy-duty cast aluminum support frame, this 50 mm diameter pulley is mounted on heavy-duty, low friction cone bearings. Fits table tops up to 1.75" thick (4.5 cm).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty