Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Distilling Flask, Borosilicate Glass, United Scientific Supplies
Supplier: United Scientific Supplies
FG4620-2000 FG4620-100 FG4620-250 FG4620-500 FG4620-125 FG4620-1000
76531-720PK 94.03 USD InStock
76531-720 76531-722 76531-718 470339-484 470339-050 76531-716
Distilling Flask, Borosilicate Glass, United Scientific Supplies
Flasks Distillation Flasks
Flasks with a long side arm for distillation applications.
  • Features large white block lettering for easy identification of capacity
Made from low-expansion borosilicate glass, these flasks feature uniform wall thickness for superior mechanical strength and shock resistance.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty