Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Disposable Plastic Tweezers

Supplier: United Scientific Supplies
UNPTWZR-NS
470339-496EA 27.49 USD InStock
470339-496
Disposable Plastic Tweezers
Forceps
Easy grip disposable plastic tweezers.

  • Manufactured from polypropylene
  • Overall length: 15 cm
  • Tip length: 5.5 cm
  • Tip sides taper from 0.3 to 0.9 cm wide
  • Maximum jaw opening: 4.5 cm

Molded tweezers are useful for a variety of applications where handling small parts is necessary. Polypropylene construction enables autoclaving if desired. Nonsterile, packed 100 per bag.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty